Would you like to log in again?
CrossLab Autosampler Valve Supplies for HPLC, Agilent Technologies
Supplier: Agilent
CrossLab
5068-0229 0100-1852 5068-0209 5068-0232 5067-4131 1535-4045 5068-0006 5068-0099 5068-0232 5068-0209 0101-1417 0100-1852 0101-1417 1535-4045 5320-0010 5067-4131 5068-0006 5068-0229 5320-0010 5068-0099
AG5068-0232EA 709.67 USD InStock
CAAG5320-0010 CA5068-0099 CA5068-0232 AG5068-0232 CA76514-502 AG0100-1852 CA5068-0209 AG5068-0099 AG5320-0010 CA5068-0229 AG5068-0229 AG5068-0006 AG5067-4131 AG5068-0209 AG1535-4045 AG0101-1417 CAAG5068-0006 CA0101-1417 CAAG5067-4131 CA1535-4045
CrossLab Autosampler Valve Supplies for HPLC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
A full range of supplies for autosampler injection valves, including rotor seals and stator faces.
  • The rotor is a highly stressed part of autosamplers that is constantly switched back and forth
  • Rotor material and surface finish govern its durability and lifetime
  • Agilent rotors are tested to ensure the longest possible lifetime
  • Use of non-Agilent valve supplies can lead to shorter lifetime, potential carryover, and leakage
Agilent provides a full range of supplies, including rotor seals and stator faces, for different autosampler injection valves. Specific designs are available to suit all application areas.
All valve supplies are manufactured to Agilent's highest levels of quality and have been tested in Agilent instruments to ensure they work perfectly with the rest of the system to help you increase your analytical, instrument, and laboratory efficiency.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty