Would you like to log in again?
CrossLab Column Nuts for GC
Supplier: Agilent
CrossLab
5180-4109 5190-1439 5188-5312 0100-1378 5190-1437 19251-60575 0100-1378 G3510-20018 5181-8830 19243-80510 05921-21170 05988-20066 0100-1365 0100-1375 5181-1292 19256-60590 0100-1375 G3510-20018 5020-8292 5020-8293 5180-4115 8710-0510 05988-20066 3150-0417 0100-1365 19251-60575 5180-4115 19256-60590 8710-0510 5190-1438 5181-8830 5190-1437 5181-1292 5020-8292 5183-4732 18740-20800 05921-21170 5183-4732 5020-8294 18740-20800 5180-4109 5180-4113 5190-1438 5188-5312 5020-8293 19243-80510 5020-8294
AG05921-21170EA 34.96 USD InStock
CA5180-4115 AG05921-21170 CA5181-1292 AG0100-1365 CAAG5183-4732 CA76514-604 AGG3510-20018 CA76514-606 CAAG5190-1438 CAAG5190-1437 AG5190-1437 CA76514-602 AG5190-1439 AG5190-1438 CA19243-80510 CA76514-566 AG05988-20066 CAAG5188-5312 AG3150-0417 AG5180-4115 AG5188-5312 CA76508-016 AG5183-4732 AG19256-60590 AG19251-60575 CA0100-1365 CA5180-4109 CAAGG351020018 AG8710-0510 AG5180-4113 CA0100-1378 AG5020-8294 AG0100-1375 AG5020-8293 CA76514-516 AG5020-8292 AG0100-1378 AG19243-80510 AG5181-8830 AG18740-20800 CA8710-0510 AG5181-1292 AG5180-4109 CA76508-102 CA19251-60575 CA0100-1375 CA19256-60590
CrossLab Column Nuts for GC
Fittings and Connectors
Column nuts come in ¹/₁₆" stainless steel, ¹/₈" and ¹/₄" brass, and new self tightening nuts.
  • No retightening means less wasted time
  • Finger-tight design lets anyone make consistent quality connections without tools
  • Low-torque seal prevents ferrules from sticking or crumbling
  • Lower column bleed prolongs column life
One of the most basic tasks in GC, installing the column, can be challenging. The GC oven is a dark, small space, and column installation uses tiny parts that need to be assembled with precision. Installation of the column into both the inlet and detector must be done properly to get accurate and reproducible results. To facilitate good column installation, Agilent offers a selection of capillary column nuts for GC fittings. The latest innovation, the Self Tightening column nut, prevents leaking and the need to retighten when using graphite/polyimide blend ferrules, which are recommended for use with oxygen sensitive detectors such as ECD or mass spectrometry.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty