Would you like to log in again?
Sample Containment Accessories, Agilent Technologies
Supplier: Agilent
5182-0575 5067-0243 9301-0722 5182-0575 5067-0243 9301-6384 9301-6384 9301-0722
AG5182-0575EA 14.86 USD InStock
AG5182-0575 AG9301-6384 AG9301-0722 AG5067-0243 CA76515-132 CA5182-0575 CA5067-0243 CAAG9301-6384
Sample Containment Accessories, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Securely hold 2 ml vials for table top sample prep or store up to 50, 2 ml vials.
  • Agilent 12 mm vial racks are ideal for holding 2 ml glass vials ( 2 ml screw vials, 2 ml crimp vials or 2 ml snap vials) for table top sample prep
  • Vial storage boxes are able to hold 50 × 2 ml vials instead which offer a cover when compared to an open vial rack
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty