Would you like to log in again?
CrossLab Purge and Trap Supplies for GC
Supplier: Agilent
CrossLab
DY70008101 5188-1447 5188-8818 5182-0850 5182-0775 391895901 5188-8819 5188-1447 5188-8818 DY50573990 5188-8816 DY50573990 5182-0850 5182-0775 5182-0847 DY70007791 5182-0847 5188-8816 392607912 5188-8817 5188-8820 5182-0852 392607912 5182-0796 391895902 5182-0845 5190-2234 5182-0845 5190-2232 DY70007791 5182-0846 5188-8820 5182-0851 5190-2231 5182-0851 5188-8817 5182-0846 391895901 5190-2232 5188-8814 5188-8819 5182-0852 5190-2231 5182-0849 5182-0849 5188-8814 5190-2234 5182-0796 391895902
AGDY70008101PK 163.01 USD InStock
CAAG5182-0847 AGDY70008101 CAAG5182-0845 CAAG5182-0846 AG5182-0847 AG5182-0849 AG5182-0845 AG5182-0846 CAAG391895902 AG391895901 CAAG391895901 AGDY50573990 AG391895902 AG5188-1447 CAAG5190-2234 AG5182-0796 CAAG5182-0775 CAAG5190-2232 CAAG5190-2231 CAAG5188-8816 CAAG5188-8817 CAAG5188-8814 CAAG5182-0850 CAAG5182-0851 CAAG5188-8818 CAAG5188-8819 CAAG392607912 CAAG5182-0849 AGDY70007791 CA5182-0852 AG5188-8820 AG5182-0850 AG5182-0851 AG5182-0775 AG5182-0852 AG5190-2232 AG5190-2231 CAAG5188-1447 AG5188-8817 AG5190-2234 AG5188-8816 AG5188-8819 AG5188-8818 CAAGDY50573990 CAAG5182-0796 CAAGDY70007791 AG392607912 CA5188-8820 AG5188-8814
CrossLab Purge and Trap Supplies for GC
Chromatography Supplies Chromatography Spare Parts
Find the supplies you need for Teledyne Tekmar purge and trap instruments.
  • Increase sample throughput up to 100% to increase profitability of the GC system
  • Highest resolution with lowest carryover
  • Exceptional inertness throughout the flow path prevents loss of active, polar, and high-boiling compounds
Find supplies for the following Teledyne Tekmar purge and trap instrumentation. The Lumin purge and trap concentrator is the recommended sample preparation system for removing VOCs from various matrices. The AQUATek 100 Waters-only autosampler provides 100-vial automated sample preparation for the analysis of liquid samples. The Atomx automated VOC sample preparation system, is suitable for those analyzing soil and water samples. The Atomx combines the features of the purge and trap with those of an autosampler in one system as well as automating methanol extractions for high-level soils. All instruments are available for purchase directly from Agilent.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty