Would you like to log in again?
CrossLab ISIS (Integrated Sample Introduction System) Supplies for ICP-MS (7700/8800), Agilent Technologies
Supplier: Agilent
CrossLab
5182-2581 G3138-65119 G3138-65122 G4911-67000 G4911-60011 5043-0010 G3138-65122 G4911-68202 5043-0010 G3138-65118 5064-8029 5064-8016 G3138-65118 5064-8028 5064-8033 G3138-65119 G4911-20021 G4911-68201 G4911-20021 G4911-67000 G4911-67001 5064-8032 5064-8020 3610010100 5064-8033 G4911-68202 5064-8032 5064-8014 G4911-67001 5064-8029 G3138-65121 5064-8014 5064-8020 G4911-68201 G3138-65117 5064-8016 G4911-01201 3610010100 5182-2581 G4911-01201 G3138-65121 G3138-65117 G4911-60011 5064-8028
AGG4911-20021EA 95.8 USD InStock
AGG4911-20021 CAAGG313865118 CAAGG491101201 AGG4911-01201 CA5064-8020 CAAGG491168202 AGG3138-65117 CAAGG491168201 AGG4911-68201 AGG4911-68202 CAAGG491120021 CA5064-8028 CAAG5064-8033 CAAGG491160011 CAAG5064-8014 CAAG5182-2581 AG5064-8033 AGG3138-65118 AG5064-8032 AGG3138-65119 AG5064-8016 AG5182-2581 AG5064-8014 CAAG5064-8032 AG5043-0010 CAAGG491167000 AG3610010100 CAAG3610010100 CAAGG491167001 CA5043-0010 AGG4911-60011 AGG4911-67000 AGG3138-65121 AGG3138-65122 AGG4911-67001 CAG3138-65117 CAAG5064-8029 CA5064-8016 AG5064-8020 CAAGG313865121 CAG3138-65119 CAAGG313865122 AG5064-8029 AG5064-8028
CrossLab ISIS (Integrated Sample Introduction System) Supplies for ICP-MS (7700/8800), Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Supplies and sample loops for Agilent ISIS switching valve used with 7700 and 8800 ICP-MS.
  • Complete kit of standard consumable parts for ISIS high sample throughput discrete sampling are provided
  • Sample loop sizes to suit your application needs are available and customizable to give you complete flexibiliy and optimal sample throughput
  • Rotor seals, nuts and ferrules for valves can be provided
Adding the Integrated Sample Introduction System (ISIS) to the 7700 and 8800 ICP-MS dramatically increases sample throughput and reduces maintenance requirements for sample introduction. Designed and manufactured by Agilent, the ISIS features a robust switching valve fitted with sample loop volume to suit your analysis requirements to allow complete flexibility.
The ISIS hardware is built into the ICP-MS instrument chassis and close coupled to increase productivity through high-speed sample uptake and rapid washout. Agilent's ISIS supplies for 7700 and 8800 ICP-MS include switching valve parts, sample loops and all associated tubing and fittings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty