Would you like to log in again?
CrossLab Programmable Temperature Vaporizing (PTV) Inlet for GC
Supplier: Agilent
CrossLab
5190-1426 5182-9747 5183-2037 5183-2037 5183-2038 5182-9749 5182-9769 5182-9749 18740-60835 18740-60835 5062-3525 5183-2036 3150-0602 5182-9758 5182-9748 5183-0318 5182-9761 5182-9763 5182-9747 5190-1426 5182-9770 5182-9761 5182-9751 5182-9759 5182-9757 5182-9760 5182-9763 9301-0713 9301-0892 3150-0602 5182-9760 5188-5313 5182-9762 9301-0892 5182-9769 5182-9762 5182-9768 5182-9759 5021-7136 5021-7133 5182-9775 5183-2058 5183-2038 5182-9754 5182-9758 5182-9756 5182-9775 5182-9768 5182-9748 5188-5356 G2617-60507 5188-5356 5182-9751 G1544-80550 5021-7137 5182-9770 5182-9757 5183-0318 5182-9756 9301-0713 G1544-80550 5188-5313 5062-3525 5183-2036 5183-2058 5021-7134 G2617-60507 5182-9754
AG5182-9770PK 264.27 USD InStock
CA5183-2058 CA5182-9768 AG5182-9770 CAAG5182-9775 CA5182-9769 CAAG5182-9770 AG9301-0713 AG5182-9775 AG5183-2037 AG5183-2038 AG5183-2036 CA5182-9761 CAG1544-80550 CA5182-9763 CA76514-816 AGG2617-60507 AG5182-9768 CA76514-812 CAAG5190-1426 AG5182-9769 AG5182-9760 AG5182-9761 AG5182-9762 AG5182-9763 CA76515-136 AG3150-0602 CAAG5062-3525 CA5183-2038 CA5183-2036 CA76514-568 AG5182-9756 AG5182-9757 CAAG5182-9758 AG5182-9758 AG5182-9759 AG9301-0892 CAAG5182-9756 CAAG5182-9757 CAAG5182-9754 AG18740-60835 CA5182-9748 CA5182-9747 CAAG5182-9751 AG5182-9751 AG5183-0318 CAAG5188-5313 AG5021-7134 CAAG5188-5356 AG5021-7136 AG5182-9754 AG5021-7137 AG5188-5313 AG5188-5356 AG5021-7133 AG5183-2058 CA76515-128 AG5190-1426 AG5182-9747 AG5182-9748 AG5182-9749 CAAG3150-0602 CA5182-9759 CAAG5182-9762 CAAG5182-9760 AGG1544-80550 CAAGG261760507 CA5183-0318 AG5062-3525
CrossLab Programmable Temperature Vaporizing (PTV) Inlet for GC
Chromatography Supplies Chromatography Spare Parts
Agilent programmed temperature vaporizer inlets combine the benefits of split, splitless, and on-column inlets, resulting in less thermal degradation of labile compounds.
  • Perform large volume injections for low minimum detection limits
  • Get less thermal degradation of labile compounds
Agilent programmed temperature vaporizer inlets combine the benefits of split, splitless, and on-column inlets. The sample is usually injected into a cool liner, so syringe needle discrimination does not occur. Then the inlet temperature is increased to vaporize the sample. You then program vent times and temperature to achieve the equivalent of split or splitless transfer of sample vapors to the column. PTV injection is considered the most universal sample introduction system because of its flexibility.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty