Would you like to log in again?
Cleaning and Maintenance Supplies for GC/MS, Agilent Technologies
Supplier: Agilent
5183-2096 G7077-60566 5184-3571 G1099-20030 3162-0110 G1960-67100 G7077-60566 5184-3571 3162-0110 G1099-20030 G1960-67100 5183-2096
AG5183-2096KT 1227.71 USD InStock
AG5183-2096 CAAG5183-2096 AGG1099-20030 AG3162-0110 AG5184-3571 CAG1099-20030 CAAG5184-3571 CAAGG707760566 CAAG3162-0110 CAG1960-67100 AGG7077-60566 AGG1960-67100
Cleaning and Maintenance Supplies for GC/MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
A selection of replacement parts and cleaning supplies to keep your GC/MS running smoothly.
  • Ball, nut, and torx drivers available
  • Maintenance and source cleaning kits, and expendable supplies kits for systems using the DS102 rough pump
  • Cleaning powder for ion transfer capillary cleaning
Agilent offers a selection of replacement parts and cleaning supplies to help you keep your GC/MS running smoothly. Our range includes ion transfer capillary cleaning powder, ferrule kits, sorbent trap refills, and vacuum grease, among other products. Excessive background in the mass spectra, which can come from the GC or MS, indicates that the system requires maintenance. The source of contamination can sometimes be determined by identifying the contaminants. Some contaminants are much more likely to originate in the GC, while others are likely to originate in the MS.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty