Would you like to log in again?
CrossLab Split Liners for GC, Agilent Technologies
Supplier: Agilent Technologies
CrossLab
5183-4700 5183-4647 5183-4713 5183-4712 5183-4691 5183-4697 5183-4711 5183-4701 5183-4692 19251-60540 5183-4700 5183-4702 210-4004-5 5183-4701 210-4022-5 18740-80190 5183-4711 18740-60840 18740-60840 5183-4713 5183-4647 5183-4692 210-4022-5 5183-4691 5183-4699 5183-4712 18740-80190 5183-4702 5190-2275 19251-60540 5183-4697 210-4004-5 5190-2275 5183-4699
AG5183-4692PK 540 USD InStock
AG5183-4692 AG5183-4691 CA5183-4699 CA5183-4697 CA5183-4713 CA76514-904 CA18740-60840 CA76514-900 CA76508-160 CA210-4022-5 AG5183-4711 AG5183-4699 AG5183-4697 AG5183-4713 CA18740-80190 AG5183-4712 CA5183-4700 AG5190-2275 CAAG5190-2275 AG210-4022-5 AG18740-60840 CA5183-4702 CA76514-898 AG210-4004-5 CA76508-162 CA76508-164 CA76514-580 CA76508-028 AG5183-4700 AG19251-60540 AG18740-80190 AG5183-4647 AG5183-4702 AG5183-4701
CrossLab Split Liners for GC, Agilent Technologies
Chromatography Supplies Chromatography Inlet Liners
Agilent split liners are made to strict dimensional specifications for optimal inlet performance.
  • Feature Agilent’s exclusive deactivation;Agilent recommends part number 5183-4647 as the top split liner choice
Split injections are intended for use with a split/splitless injection port in gas chromatography analysis. Split liners tend to have a slightly smaller outer diameter to allow for gas flow. For ease of use and reproducibility, the liners have a positioning bead, a restriction to secure the position of the glass wool, and a feature to consistently self-position to the recommended height.;Agilent split liners are made to strict dimension specifications for optimal inlet performance and feature the tightest tolerances for od, id, taper and glass wool placement.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty