Would you like to log in again?
CrossLab Direct Connect Liners for GC, Agilent Technologies
Supplier: Agilent
CrossLab
G1544-80730 5183-4709 5183-4709 18740-80200 18740-80200 G1544-80700 5183-4710 5183-4710 G1544-80700 G1544-80730
AGG1544-80700EA 103.75 USD InStock
AGG1544-80700 AG18740-80200 AGG1544-80730 CAAG5183-4710 CA18740-80200 AG5183-4709 AG5183-4710 CAG1544-80700 CAG1544-80730 CA5183-4709
CrossLab Direct Connect Liners for GC, Agilent Technologies
Chromatography Supplies Chromatography Inlet Liners
Direct Connect liners provide sensitivity,maximum recovery, and minimal decomposition of active compounds for methods requiring splitless injection such as EPA 8270.
  • Press fit connection to the column
  • No sample exposure to inlet
  • Included in 8270 EPA application kit
  • Optimized hole size and placement
Agilent's Direct Connect liners provide maximum recovery and minimal decomposition of active compounds for methods requiring splitless injection, such as EPA 8270. They are best for relatively clean samples containing active analytes, such as water extracts. The liners directly connect with the column, similar to press-fit connectors, to aid complete transfer of sample onto the column, eliminating the problem of inlet discrimination and further increasing sensitivity. Direct connect liners are not designed for the purposes of high volume injections.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty