Would you like to log in again?
Inlet Convenience Kits for GC, Agilent Technologies
Supplier: Agilent
CrossLab
5188-6493 G1544-60585 5188-6496 5188-6497 5188-6493 G1544-60585 5188-6495 5188-6498 5188-6494 5188-6495 5188-6497 5188-6498 G3452-60730 G3452-60730 5188-6494 5188-6496
AGG3452-60730EA 551.24 USD InStock
AGG3452-60730 CA5188-6495 CA5188-6496 CA5188-6497 CA5188-6493 AG5188-6494 AG5188-6495 AG5188-6496 AG5188-6497 CAAG5188-6498 AG5188-6493 CA76515-244 AGG1544-60585 AG5188-6498 CAG3452-60730 CAAG5188-6494
Inlet Convenience Kits for GC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Convenience kits are an easy way to get all the supplies you need using one part number.
  • Designed, tested and manufactured by Agilent engineers to work perfectly with our instruments superior performance
Convenience kits are an easy way to get all the supplies you need using one part number.
Agilent's new PM kits include septa, liners, O-rings, gold seals and traps.
Delivery information: Agilent's new PM kits include septa, liners, O-rings, gold seals and traps.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty