Would you like to log in again?
CrossLab Color Coded Syringes for Manual Injection Valves, Agilent Technologies
Supplier: Agilent
CrossLab
5190-1485 5190-1485 5190-1520 5190-1515 5190-1508 5190-1480 5190-1512 5190-1501 5190-1526 5190-1508 5190-1522 5190-1526 5190-1501 5190-1505 5190-1515 5190-1480 5190-1522 5190-1512 5190-1492 5190-1499 5190-1484 5190-1484 5190-1520 5190-1494 5190-1505 5190-1499 5190-1492 5190-1494
AG5190-1492EA 86.2 USD InStock
AG5190-1492 CAAG5190-1494 AG5190-1494 CAAG5190-1492 AG5190-1499 AG5190-1512 CAAG5190-1515 CAAG5190-1512 AG5190-1515 CAAG5190-1499 AG5190-1508 CAAG5190-1508 CAAG5190-1485 CAAG5190-1484 AG5190-1484 CA5190-1501 AG5190-1485 CAAG5190-1480 AG5190-1520 AG5190-1501 CAAG5190-1505 CAAG5190-1526 AG5190-1522 AG5190-1505 CAAG5190-1522 AG5190-1526 CAAG5190-1520 AG5190-1480
CrossLab Color Coded Syringes for Manual Injection Valves, Agilent Technologies
Syringes Syringes for Chromatography
Many Agilent manual syringes for GC have color-coded barrels to facilitate volume selection.
  • The blunt-tip point style needle is designed to prevent damaging the valves internal parts
  • Can be used with any type/brand of manual injection valve
  • For fast, easy identification, each denomination (0.5 µl to 50 ml) features a distinct color boldly embossed on the glass barrel
  • Each syringe is designed with an easy-to-read volume scale and eco-friendly packaging that doubles as a syringe stand
For more help choosing the right syringe for your application, check out our syringe selection tool.
Certifications: Each syringe comes with a Certificate of Conformance, which is available online as well.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty