Find the supplies you need for Teledyne Tekmar purge and trap instruments.
- Increase sample throughput up to 100% to increase profitability of the GC system
- Highest resolution with lowest carryover
- Exceptional inertness throughout the flow path prevents loss of active, polar, and high-boiling compounds
Find supplies for the following Teledyne Tekmar purge and trap instrumentation. The Lumin purge and trap concentrator is the recommended sample preparation system for removing VOCs from various matrices. The AQUATek 100 Waters-only autosampler provides 100-vial automated sample preparation for the analysis of liquid samples. The Atomx automated VOC sample preparation system, is suitable for those analyzing soil and water samples. The Atomx combines the features of the purge and trap with those of an autosampler in one system as well as automating methanol extractions for high-level soils. All instruments are available for purchase directly from Agilent.