Would you like to log in again?
Agilent Bio MAb, Agilent Technologies
Supplier: Agilent Technologies
5190-2401 5190-2406 5190-2405 5190-2401 5190-2406 5190-2411 5190-2402 5190-2415 5190-2405 5190-2411 5190-2402 5190-2415
AG5190-2401EA 1861 USD InStock
AG5190-2401 AG5190-2411 CAAG5190-2406 AG5190-2402 CAAG5190-2405 AG5190-2405 CAAG5190-2415 AG5190-2415 AG5190-2406 CAAG5190-2402 CAAG5190-2401 CAAG5190-2411
Agilent Bio MAb, Agilent Technologies
Chromatography Columns
High density non-porous weak cation exchanger for mAb charge variant analysis.
  • A packing support composed of a rigid, spherical, highly cross-linked polystyrene divinylbenzene (PS/DVB) non-porous bead
  • Particles grafted with a hydrophilic, polymeric layer, virtually eliminating non-specific binding of antibody proteins
  • A different process is used to layer the weak cation-exchange phase to the particle making it a higher density than the Agilent Bio WCX column particles
  • Specifically designed for the separation of charge isoforms of monoclonal antibodies
Thorough characterization of monoclonal antibodies includes the identification and monitoring of acidic and basic isoforms. The Agilent Bio MAb HPLC columns feature a unique resin specifically designed for high-resolution charge-based separations of monoclonal antibodies. Bio MAb columns are available in 1.7 μm, 3 μm, 5 μm, and 10 μm sizes, providing higher resolution with the smaller particles. The Bio MAb column features a highly uniform, densely packed, weak cation exchange layer. This layer of the Bio MAb column is chemically bonded to the hydrophilic, polymeric coating of the Bio MAb column. View our full range of bioHPLC columns for your area of interest, get resources, and take advantage of special offers.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty