High resolution, non-porous ion exchange separations of proteins, peptides and other biological molecules.
- Highly crosslinked and rigid nonporous poly(styrene divinylbenzene) (PS/DVB) particles are grafted with a hydrophilic, polymeric layer, eliminating nonspecific binding
- Uniform, densely packed ion-exchange functional groups are chemically bonded to the hydrophilic layer (multiple ion-exchange groups per anchoring) to increase column capacity
Agilent Bio Ion-Exchange HPLC columns are packed with polymeric, nonporous, ion-exchange particles. Bio Ion-Exchange Columns are designed for high resolution, high recovery and highly efficient separations of peptides, oligonucleotides and proteins. The Bio IEX family offers strong cation-exchange (SCX), weak cation-exchange (WCX), strong anion-exchange (SAX) and weak anion-exchange (WAX) phases. All Bio Ion-Exchange phases are available in 1.7, 3, 5 and 10 μm non-porous particles sizes. View our full range of bioHPLC columns for your area of interest, get resources, and take advantage of special offers.