Would you like to log in again?
Agilent Bio IEX
Supplier: Agilent Technologies
5190-2467 5190-2493 5190-2460 5190-2441 5190-2466 5190-2488 5190-2460 5190-2426 5190-2461 5190-6876 5190-2488 5190-2476 5190-2439 5190-2491 5190-2483 5190-2468 5190-6882 5190-2484 5190-2491 5190-2487 5190-2485 5190-6882 5190-2448 5190-2485 5190-2439 5190-2427 5190-2476 5190-2428 5190-2481 5190-2427 5190-6876 5190-2442 5190-6875 5190-2423 5190-2471 5190-2441 5190-2466 5190-2484 5190-2425 5190-2445 5190-6875 5190-2442 5190-2483 5190-2425 5190-2423 5190-2468 5190-2428 5190-2472 5190-2493 5190-2487 5190-2445 5190-2426 5190-2471 5190-2448 5190-2481 5190-2461 5190-2467 5190-2472
AG5190-2461EA 1861 USD InStock
AG5190-2461 AG5190-2460 CAAG5190-2461 CAAG5190-2460 AG5190-2423 AG5190-2467 AG5190-2466 AG5190-2425 AG5190-2468 CAAG5190-2428 AG5190-2427 CAAG5190-2427 AG5190-2426 CAAG5190-2426 CAAG5190-2425 CAAG5190-2468 AG5190-2428 CAAG5190-2467 CAAG5190-2423 CAAG5190-2466 CAAG5190-2476 AG5190-2493 CAAG5190-2472 CAAG5190-2471 CAAG5190-2439 CAAG5190-6876 CAAG5190-6875 AG5190-2491 AG5190-2483 CAAG5190-2487 CAAG5190-2442 CAAG5190-2441 CAAG5190-2485 AG5190-2485 AG5190-2441 CAAG5190-2484 AG5190-6882 AG5190-2484 CAAG5190-2483 AG5190-2487 AG5190-2442 CAAG5190-2481 AG5190-2445 AG5190-2488 CAAG5190-6882 AG5190-2448 CAAG5190-2448 CAAG5190-2445 CAAG5190-2488 AG5190-2481 AG5190-2472 AG5190-2471 AG5190-2476 CAAG5190-2493 AG5190-6876 CAAG5190-2491 AG5190-6875 AG5190-2439
Agilent Bio IEX
Chromatography Columns
High resolution, non-porous ion exchange separations of proteins, peptides and other biological molecules.
  • Highly crosslinked and rigid nonporous poly(styrene divinylbenzene) (PS/DVB) particles are grafted with a hydrophilic, polymeric layer, eliminating nonspecific binding
  • Uniform, densely packed ion-exchange functional groups are chemically bonded to the hydrophilic layer (multiple ion-exchange groups per anchoring) to increase column capacity
Agilent Bio Ion-Exchange HPLC columns are packed with polymeric, nonporous, ion-exchange particles. Bio Ion-Exchange Columns are designed for high resolution, high recovery and highly efficient separations of peptides, oligonucleotides and proteins. The Bio IEX family offers strong cation-exchange (SCX), weak cation-exchange (WCX), strong anion-exchange (SAX) and weak anion-exchange (WAX) phases. All Bio Ion-Exchange phases are available in 1.7, 3, 5 and 10 μm non-porous particles sizes. View our full range of bioHPLC columns for your area of interest, get resources, and take advantage of special offers.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty