Would you like to log in again?
Agilent Bio SEC-5, Agilent Technologies
Supplier: Agilent
5190-2536 5190-2527 5190-2540 5190-2535 5190-2545 5190-6861 5190-2528 5190-2544 5190-6860 5190-6862 5190-6862 5190-2518 5190-2535 5190-2544 5190-6860 5190-2529 5190-2527 5190-2536 5190-2529 5190-6861 5190-2526 5190-2518 5190-2545 5190-2528 5190-2526 5190-2523 5190-2540 5190-2523
AG5190-6862EA 761.04 USD InStock
AG5190-6862 CAAG5190-2540 AG5190-6861 AG5190-2540 AG5190-6860 AG5190-2544 CAAG5190-2528 CAAG5190-2527 AG5190-2545 AG5190-2523 CAAG5190-6861 AG5190-2526 CAAG5190-2526 CAAG5190-6860 AG5190-2528 CAAG5190-6862 AG5190-2527 CAAG5190-2545 CAAG5190-2523 CAAG5190-2544 AG5190-2529 CAAG5190-2518 AG5190-2535 CAAG5190-2536 AG5190-2536 CAAG5190-2535 AG5190-2518 CAAG5190-2529
Agilent Bio SEC-5, Agilent Technologies
Chromatography Columns
Widest pore size range SEC columns, from 100 to 2000 Å, with low operating pressures.
  • Maximum recovery for a broad range of size-based, biomolecule separations
  • Outstanding reproducibility and column lifetime
  • Excellent stability, even under high pH, high salt, and low salt conditions
  • Compatible with most aqueous buffers
Agilent Bio SEC-5 HPLC columns are packed with 5 μm silica particles coated with a proprietary, neutral, hydrophilic layer for maximum efficiency and stability. Our specially designed packing also promotes high pore volume, improving both peak capacity and resolution. Agilent Bio SEC-5 HPLC columns are available in 100, 150, 300, 500, 1000 and 2000 Å. View our full range of bioHPLC columns for your area of interest, get resources, and take advantage of special offers.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty