Would you like to log in again?
CrossLab Ultra Inert Liners for GC, Agilent Technologies
Supplier: Agilent
CrossLab
5190-7020 5190-3983 5190-7045 5190-2294 5190-3170 5190-7013 5190-3164 5190-3167 5190-7020 5190-2295 5190-3169 5190-5105 5190-7012 5190-7013 5190-4047 5190-5112-005 5190-3166 5190-5112 5190-7014 5190-3165 5190-2295 5190-4056 5190-4056 5190-2293 5190-3173 5190-3172 5190-3166 5190-3162 5190-3169 5190-5112-005 5190-3171 5190-7045 5190-5105-005 5190-3162 5190-2293 5190-3167 5190-5112 5190-2292 5190-4048 5190-3168 5190-3164 5190-3165 5190-4007 5190-2294 5190-3170 5190-7011 5190-3171 5190-4007 5190-7012 5190-3173 5190-4048 5190-4047 5190-5105-005 5190-5105-025 5190-7014 5190-5105 5190-3163 5190-3163 5190-3168 5190-2292 5190-6168 5190-3983 5190-3172 5190-6168
AG5190-4047EA 45.93 USD InStock
AG5190-4047 CA51905105005 AG5190-4007 AG5190-4048 CA5190-2293 CA5190-2294 CA5190-7020 CA5190-2292 CA5190-2295 CA5190-3983 AG5190-2295 AG5190-3983 CAAG5190-7045 CAAG5190-5105 CA5190-4047 CA5190-4007 AG5190-2292 AG5190-2294 AG5190-7020 AG5190-2293 CA5190-4048 AG5190-5112 AG5190-6168 AG5190-7014 CAAG5190-3173 CAAG5190-7013 CAAG5190-7012 CAAG5190-7014 CA5190-3162 CA5190-3163 CA5190-3166 CA5190-3167 AG5190-3170 AG5190-7011 CAAG5190-3170 CA5190-4056 CA5190-3164 AG5190-7012 AG5190-7013 CA5190-3165 AG51905105-025 AG5190-3173 AG5190-3171 CA5190-3168 CA5190-3169 AG5190-3172 AG5190-3166 AG5190-3167 AG5190-3164 AG5190-3165 AG5190-4056 AG5190-5105 AG5190-3168 AG5190-3169 AG5190-5112005 CA51905112005 CA5190-3171 CA5190-3172 AG5190-7045 CA5190-5112 AG5190-3162 AG5190-5105005 AG5190-3163 CA5190-6168
CrossLab Ultra Inert Liners for GC, Agilent Technologies
Chromatography Supplies Chromatography Inlet Liners
Agilent Ultra Inert inlet liners prevent adsorption and ensures accurate sample transfer onto the GC column.
  • Glass-fritted liners now available for splitless and universal applications to help increase reproducibility and extend liner life
  • Consistent high level of inertness for greater sensitivity, accuracy, and reproducibility
  • Ultra Inert split and splitless liners, with and without wool, for a wide variety of applications
  • Liners with inert wool retain nonvolatiles, making trace-level analysis of difficult active compounds easier
  • Unique touchless packaging with precleaned, chromatographically ready O-ring minimizes the risk of contamination from touching
Inlet liners facilitate sample vaporization in the GC inlet and are offered in variety internal volumes with unique features to suit specific applications. Ultra Inert liners are treated with Agilent’s exclusive deactivation to ensure maximum inertness.
Agilent Ultra Inert inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace-level analysis of active analytes. Liners with inert wool retain nonvolatiles, prolonging column life and lowering frequency of MS source maintenance.
Compatible with Agilent GC systems, including Intuvo 9000, 7890, 6890, and 5890. Also available for GC systems from other manufacturers.
Certifications: Certificate of Performance - each Ultra Inert liner deactivation lot is certified to ensure efficient, consistent coverage using both acidic and basic probes at trace levels (2 ng) on-column.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty