Would you like to log in again?
Interface Cone Supplies for ICP-MS
Supplier: Agilent
CrossLab
G3280-01507 G1820-65025 5190-9614 G3280-67009 G3280-60502 5190-9614 G3280-01507 9300-2574 G3280-60502 G3280-67009 9300-2574
AG5190-9614EA 236.55 USD InStock
AG5190-9614 AG9300-2574 AGG1820-65025 CAAGG328001507 AGG3280-60502 CAG3280-67009 CA5190-9614 CAAGG328060502 CA9300-2574 AGG3280-67009 AGG3280-01507
Interface Cone Supplies for ICP-MS
Chromatography Supplies Chromatography Spare Parts
Accessories and supplies for skimmer and sampling cones for use with our ICP-MS portfolio.
  • Graphite gasket for sampling cone that seals the sampling cone to the vacuum chamber interface
  • Available interface wrenches (in order to remove and install the skimmer cone) and sampling cone wrenches (used to remove the sampling cone retaining ring)
  • Cone cleaning and maintenance supplies including fixtures, cone magnifier, cotton swabs, Citranox and Alumina powder
Agilent's high quality ICP-MS systems contain interfaces that have a step-down vacuum stage, located between a pair of conical metal plates, known as interface cones. In common terminology, the first and second cones are referred to as the sample cone and skimmer cone, respectively. Agilent has all your interface cone maintenance needs covered with our range of supplies and accessories.
A range of accessories, components and spare parts are available including cone tools, magnifier and cleaning fixtures, sampling cone carbon/graphite gaskets, cotton cleaning swabs, Citranox and Alumina powder.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty