Would you like to log in again?
CrossLab SPME Fibers, Agilent Technologies
Supplier: Agilent
CrossLab
5191-5879 5191-5871 5191-5875 5191-5870 5191-5875 5191-5872 5191-5876 5191-5879 5191-5871 5191-5870 5191-5878 5191-5876 5191-5874 5191-5872 5191-5873 5191-5874 5191-5878 5191-5873
AG5191-5870PK 649.41 USD InStock
CA5191-5874 AG5191-5870 AG5191-5879 CAAG5191-5878 CAAG5191-5879 AG5191-5871 AG5191-5872 CAAG5191-5873 CAAG5191-5876 AG5191-5873 CAAG5191-5875 AG5191-5874 CAAG5191-5870 AG5191-5875 AG5191-5876 CAAG5191-5872 CAAG5191-5871 AG5191-5878
CrossLab SPME Fibers, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
SPME fibers are available in a variety of chemistries and formats.
  • SPME fibers can be used multiple times when treated with the proper care and caution
  • Each fiber has a color-coded or notched hub, indicating the type of coating on the fiber
  • Fiber kits contain only the fibers. For a first-time order, you will also need to order the appropriate holder for your needs
Agilent offers SPME fibers in a range of chemistries, formats, and for use with autosamplers or manual injections. SPME fibers can be used multiple times as long as they are treated with the proper care and caution. Each fiber has a color-coded or notched hub, indicating the type of coating on the fiber.
Delivery information: Fiber kits contain only the fibers. For a first-time order, please also order the appropriate holder for your needs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty