Would you like to log in again?
SPME Fiber Accessories, Agilent Technologies
Supplier: Agilent Technologies
5191-5877 392609902 392609902 5191-5877
AG392609902EA 407 USD InStock
CAAG392609902 AG392609902 CAAG5191-5877 AG5191-5877
SPME Fiber Accessories, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Extra parts to assist in SPME sample preparation.
  • SPME fiber kits contain only the fibers
For a first-time order, you will also need to order the appropriate fiber holder for your application needs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty