Would you like to log in again?
CrossLab Flame Ionization Detector (FID) for GC, Agilent Technologies
Supplier: Agilent
CrossLab
19301-60660 8710-1346 5200-0176 G3431-60680 19231-60680 19231-80530 G1531-67000 9301-0985 19231-80520 5180-4153 5180-4150 19231-21050 394958700 G1531-60690 G1531-21090 19231-21050 5181-3311 G1531-67001 19231-21060 8710-1346 G1531-60680 8710-1561 394958700 5080-4978 5200-0178 G1531-67000 5180-4152 8710-1561 19231-20940 19301-60660 G1531-20700 5200-0177 G1531-20700 G3431-60680 19231-20910 19244-80610 G1531-60690 G4591-60041 5180-4150 19231-60680 G1531-20690 19231-20940 G1531-20690 5200-0177 5180-4153 5181-3311 19231-21060 5200-0176 G1531-21090 19231-80520 19231-80530 G4591-60041 19244-80610 5200-0178 5080-4978 G1531-60680 G1531-67001 5180-4152 19231-20910 9301-0985
AG5180-4152PK 45.23 USD InStock
AG5180-4152 AGG1531-21090 AG5180-4153 AG5180-4150 AG19244-80610 AG5200-0177 AG5181-3311 AG5200-0176 AG5200-0178 AG19231-20910 AGG1531-67001 CAG1531-21090 AG8710-1346 AGG1531-60690 AGG1531-67000 AG19301-60660 AG19231-80530 CA19231-20910 CAAG1923180530 AGG1531-20700 AG9301-0985 CA8710-1561 CA76508-024 AG19231-21050 CA5200-0176 CAAG1923121050 CA5200-0177 CAAGG459160041 AGG1531-60680 CA19231-80520 CA5200-0178 AG394958700 AGG3431-60680 CAG1531-60680 CAAG394958700 CA5180-4152 CA5181-3311 CA5180-4153 CAAG5080-4978 CA5180-4150 AG5080-4978 AG19231-21060 AGG1531-20690 AG19231-80520 AG8710-1561 CA19301-60660 CAAG1923120940 CA76515-240 CA19244-80610 AG19231-60680 CA9301-0985 CAG3431-60680 CA8710-1346 CAG1531-67001 AG19231-20940 CA19231-21060 AGG4591-60041 CAG1531-67000 CA76515-238 CA76515-236
CrossLab Flame Ionization Detector (FID) for GC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Agilent provides a full range of supplies required to run your FID.
  • New FID jets include a shorted jet tail with a wider inner diameter to facilitate column installation
  • Etched rings on jet heads help to easily identify the orifice size of each jet
  • The newly designed FID jets are compatible with both capillary and packed columns, eliminating the need to switch parts and reducing your downtime;The most popular and versatile detector
  • Designed for maximum sensitivity and ease-of- use
  • Flame can be ignited from keyboard or automatically
Flame ionization detection is the most commonly used GC detection method due to it's superior ability to measure hydrocarbons. Using a carrier gas, the sample is ionized via a hydrogen-air flame, then collected using a polarizing voltage. The current produced is measured and used to quantify the amount of sample being burned. Agilent provides a full range of supplies required to run fid chromatography. This includes igniter glow plugs, collector nuts, cleaning and maintence kits, and our newly designed single-piece FID jets. These innovative jets are aimed to improve ease of installation by eliminating damage to the threads as well as your column heads and improve your overall fid analysis.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty