Would you like to log in again?
Smart SPME Fibers, Agilent Technologies
Supplier: Agilent
5610-5878 5610-5876 5610-5870 5610-5872 5610-5871 5610-5878 5610-5870 5610-5873 5610-5873 5610-5874 5610-5874 5610-5879 5610-5872 5610-5879 5610-5875 5610-5876 5610-5875 5610-5871
AG5610-5874PK 858.45 USD InStock
AG5610-5874 CAAG5610-5878 AG5610-5873 AG5610-5872 CAAG5610-5879 AG5610-5871 AG5610-5870 CAAG5610-5873 CAAG5610-5874 CAAG5610-5875 CAAG5610-5876 CAAG5610-5870 CAAG5610-5871 CAAG5610-5872 AG5610-5879 AG5610-5878 AG5610-5876 AG5610-5875
Smart SPME Fibers, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Smart SPME fibers for PAL3 instrumentation.
  • Smart technology enables critical parameter tracking for each unique SPME fiber
  • Automatic application of the correct parameters for the individual smart SPME fiber
  • Compatible with all PAL3 models and existing SPME tools
  • Color coded to indicate the fiber coating and film thickness
  • Preinstalled with holder for easy maintenance
  • Smart SPME fibers kits available for method development
Agilent offers smart SPME fibers for compatibility with all PAL3 models including PAL 3 Series II smart instrumentation. Each smart SPME fiber is equipped with a unique smart chip containing parameters, ranges, and usage history for the specific fiber. Agilent smart SPME fibers are color coded to easily identify chemistry and thickness. Whether you are a new SPME user or switching to smart technology, you can be confident that performance of Agilent smart SPME fibers remains of the highest quality.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty