Would you like to log in again?
HC-C18(2) & TC-C18(2), Agilent Technologies
Supplier: Agilent
Agilent
588915-902 588925-902 588935-902 588915-902 588925-902 588935-902 520518-904 520518-904
AG588925-902EA 451.55 USD InStock
CAAG588915-902 CAAG588925-902 AG588925-902 CAAG520518-904 CAAG588935-902 AG520518-904 AG588915-902 AG588935-902
HC-C18(2) & TC-C18(2), Agilent Technologies
Chromatography Columns
Low-cost C18 columns for legacy HPLC applications. Available in moderate and high carbon loads.
  • Economical choice for simple separations, where speed is not a concern
  • Available in high carbon load and moderate carbon load bonding
Agilent HC-C18(2) and TC-C18(2) columns are an economical choice for simple separations, where speed is not a concern. Agilent HC-C18(2) is a more retentive C18 with a higher carbon load for more complex samples. Agilent TC-C18(2) is bonded to moderate density for superior resolution of mixed polar and nonpolar samples.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty