Would you like to log in again?
CrossLab Graphite Furnace AAS Graphite Electrodes, Agilent Technologies
Supplier: Agilent
CrossLab
6310001600 6310001700 6310001700 6310003500 6310003500 6310003400 6310003400 6310001600
AG6310003400EA 346.13 USD InStock
CAAG6310003500 AG6310003400 CAAG6310003400 CAAG6310001600 AG6310001600 AG6310003500 CAAG6310001700 AG6310001700
CrossLab Graphite Furnace AAS Graphite Electrodes, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Spare graphite furnace electrodes for optimum performance with maximum tube life.
  • Keeping spare electrodes on-hand helps you maximize uptime
  • Good electrical contact ensures optimum performance and maximizes tube life
  • High purity graphite reduces spurious absorption from trace contaminants and improves signal-to-noise ratio
Agilent graphite furnace AAS electrodes provide good electrical contact, ensuring you achieve optimum performance with maximum tube life.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty