Would you like to log in again?
AdvanceBio Glycan Mapping, Agilent Technologies
Supplier: Agilent
651750-913 651750-913 683975-913 685775-913 859700-913 821725-905 821725-906 821725-905 859700-913 821725-906 683975-913 685775-913
AG821725-906PK 835.11 USD InStock
CAAG683975-913 CAAG821725-905 AG821725-906 AG683975-913 CAAG821725-906 AG821725-905 AG651750-913 AG685775-913 CAAG685775-913 CAAG859700-913 CAAG651750-913 AG859700-913
AdvanceBio Glycan Mapping, Agilent Technologies
Chromatography Columns
AdvanceBio Glycan Mapping Columns for fast, reproducible, high resolution glycan characterization.
  • Speed: Obtain glycan maps in less than 10 minutes, superior results - in 40% less time than the competition
  • High-resolution: Characterize both labeled and unlabeled glycans, and identify any glycosylation changes that occur due to process variables
  • Flexibility: 2.7 µm superficially porous particles for high resolution with less backpressure. For UHPLC analysis 1.8 µm particles are an ideal choice
  • Reproducibility: QA tested with a 2-AB labeled IgG N-linked glycan sample to ensure consistent quality and performance
AdvanceBio Glycan Mapping columns are designed and manufactured to deliver fast, high resolution, reproducible glycan identification using HILIC chromatography. Two UHPLC configurations are available: 2.7 µm superficially porous, for high resolution and lower backpressure, and 1.8 µm for highest resolution.
AdvanceBio Glycan Mapping columns leverage technology that optimizes results for MS and fluorescence detection. They can deliver glycan maps in less than 10 minutes - 40% less time than competing products - and can be ordered with a selection of standards for performance testing and retention mapping of labeled and unlabeled glycans.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty