Would you like to log in again?
Autosampler Sample Vials, Beakers and Adapters, Agilent Technologies
Supplier: Agilent
6610008200 9910116900 9910028200 6610008200 9910028200 9910115600 6610025900 6610025900 9910115600 9910116900
AG9910116900PK 215.23 USD InStock
CA6610025900 AG9910116900 AG9910028200 CA9910028200 AG6610025900 CAAG9910115600 AG9910115600 CAAG9910116900 AG6610008200 CAAG6610008200
Autosampler Sample Vials, Beakers and Adapters, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Sample vials, beakers and adapters for use with Agilent PSD 100/120 Programmable sample dispenser.
  • Sample vials are of heavy-duty construction to prevent cracking or tipping
  • The conical design of the vials allows for use of nearly the entire sample
  • Sample vials are molded from translucent, LDPE and are recommended for use with most aqueous solutions
  • There is a choice of vials available for either the standard 50 position carousel (2 ml capacity) or the optional 130 sample high capacity carousel (1.1 ml capacity)
  • Larger volume reagent containers are suitable for holding bulk quantities of blank solution for dilution and standard preparation, bulk standard for standard preparation, other reagent solutions and chemical or matrix modifiers
Agilent provides a range of supplies for the PSD 100/120 Programmable sample dispenser used with Agilent GFAAS systems, including sample vials, beakers, adapters, dispensing capillary assemblies, dispensing syringe and replacement plungers.
Adapters are also available to enable smaller sample vials to be located in the larger carousel positions when working with smaller volumes of chemical modifiers or other reagents.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty