Would you like to log in again?
Flow Through Cells for UV-Vis and UV-Vis-NIR Systems, Agilent Technologies
Supplier: Agilent Technologies
6610012600 6610010100 5063-6570 5063-6570 6610010000 5062-2476 6610019900 5188-8003 5188-8003 5061-3396 6610020100 5063-6572 6610014200 5188-8005 6610014100 5063-6573 5062-2476 5188-8005 6610020000 0100-1225 5063-6573 5061-3397 5063-6572 5065-9918 5061-3396 6610020000 6610008900 6610012600 5188-8004 5061-3397 6610015200 5061-3398 0100-1225 6610010000 5065-9918 5188-8004 6610008900 6610020200 6610014200 6610014100 6610015200 6610019900 5061-3398 6610020200 6610020100 6610010100
AG0100-1225PK 1682 USD InStock
CA0100-1225 CAAG6610019900 CAAG5063-6572 AG0100-1225 CAAG5063-6570 AG6610015200 CAAG5188-8005 AG5188-8003 CAAG5188-8003 CAAG5188-8004 AG5188-8004 CA5061-3397 CA5061-3396 CAAG6610012600 CAAG6610008900 AG6610019900 CAAG6610010000 CAAG5065-9918 CAAG6610010100 CA5061-3398 CAAG6610014200 AG5188-8005 CA5063-6573 AG5065-9918 CAAG6610014100 AG5061-3398 AG6610012600 AG6610008900 AG6610010000 AG6610014200 AG6610010100 CAAG6610020100 AG6610014100 CAAG6610020200 CAAG6610020000 AG6610020100 AG6610020200 AG5063-6573 AG5062-2476 AG5063-6572 AG5063-6570 AG5061-3397 AG5061-3396 AG6610020000 CAAG5062-2476 CAAG6610015200
Flow Through Cells for UV-Vis and UV-Vis-NIR Systems, Agilent Technologies
Cuvettes Spectrophotometer Cuvettes
Flow thru cells with 0.1 - 100 mm path length, choose aperture and 15/20 mm z height for 8454/Cary UV.
  • Flow through cells are designed for use with a sipper to reduce sample measurement time and operator-induced errors associated with manually filling cuvettes
  • 10 mm pathlength flow through cells fit any standard cell holder
  • Flow through cells feature non transmitting, self masking black walls which improve sensitivity by eliminating stray light from measurements
  • Exceptional accuracy in terms of optical path length and parallelism
  • Unique surface flatness of the optical windows
  • High temperature stability and high chemical resistance
  • Outstanding measurement reproducibility
  • Guaranteed transmission of >80% from 200-3500 nm (depending on the cell window material)
  • Flow-through cells do not include tubing/fittings (unless indicated otherwise)
Flow through cells allow the sample to pass through the cell and are connected to the sample source via tubing. They require a pump such as a sipper accessory, to draw sample into the cell for measurement and out again afterwards. This improves productivity by reducing the sample measurement time and operator-induced errors associated with manually filling cuvettes.
Agilent's oval aperture cells combine low volume with excellent flow characteristics, and are recommended for automated analyses such as dissolution testing. Long path length flow cells are useful for low concentration samples and require the long path length rectangular cell holder.
Ordering information: Flow-through cells do not include tubing/fittings (unless indicated otherwise).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty