Would you like to log in again?
CrossLab Argon Gas and Air Inlet Filters for ICP-OES, Agilent Technologies
Supplier: Agilent
CrossLab
G8000-68002 6810008600 6810008700 6810008600 6810008700 G8010-60189 G8000-68002 G8010-60189 G8020-68016 G8020-68016
AGG8000-68002EA 169.61 USD InStock
AGG8000-68002 AGG8020-68016 CAAG6810008600 AGG8010-60189 AG6810008700 CAG8000-68002 CAG8020-68016 AG6810008600 CAAG6810008700 CAAGG801060189
CrossLab Argon Gas and Air Inlet Filters for ICP-OES, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Safety and preventative measure assure the highest purity gas/air to the ICP-OES instrumentation.
  • Delivers clean gases for accurate analyses.
  • Both inlet and outlet connectors are equipped with stainless steel frits to prevent particulates from entering the gas stream.
  • Air inlet filter captures laboratory dust and debris to protect and prolong the instrument.
  • Easily replaced.
Agilent high-performance gas purifiers are designed to optimize ICP-OES system performance. Cartridges remove impurity gases, moisture, and hydrocarbons from ICP-OES gas supplies. The ICP-OES air inlet filter protects the instrument from debris and particles in the lab environment from entering the system. Replacement reminder tags ensure sensitivity and results aren’t impacted for optimal sensitivity and results. Agilent gas purifying cartridges for ICP-OES are a cost-effective safety and preventative measure to assure the supply of the highest purity gas to your ICP-OES instrumentation. Air inlet filter captures laboratory dust and debris to protect and prolong the instrument.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty