Would you like to log in again?
O-Rings for Spray Chambers, Agilent Technologies
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CrossLab
6910010400 6910012100 6910005200 9910093400 6910013600 6910013600 6910036800 6910036900 6910009700 6910037000 6910000100 9910093500 9910045700 6910015200 6910004000 6910036900 9910026500 6910004000 6910037000 9910093500 6910010400 6910009800 6910017400 6910012100 6910005200 9910045700 6910017400 9910026500 6910015200 9910093400 6910000100 6910036800 6910009800 6910009700
AG9910026500KT 139 USD InStock
AG9910026500 CA6910012100 CAAG6910036800 CA6910037000 CAAG6910036900 AG6910005200 CA9910093400 CAAG6910015200 AG9910093400 AG6910009800 CAAG6910017400 AG9910045700 AG6910009700 AG6910036800 AG6910013600 AG6910036900 AG9910093500 AG6910017400 AG6910015200 CAAG6910004000 AG6910000100 CAAG6910010400 CA6910013600 CA6910009700 AG6910004000 CA6910009800 AG6910012100 CAAG9910026500 CA6910000100 AG6910010400 CAAG9910093500 CAAG9910045700 AG6910037000 CAAG6910005200
O-Rings for Spray Chambers, Agilent Technologies
Seals Sealing Rings
O-rings for routine maintenance of the current Mark seven or earlier Mark VI flame atomization system.
  • Regular inspection of the O-rings in the flame atomization system is recommended
  • O-rings should be replaced if they are perished, show excessive wear or have any nicks or cuts on the surface
  • The regular black O-rings are suitable for use with aqueous solutuions, including acid digests
  • For compatibility with organic solvents, perfluoroelastomer (white) O-rings should be fitted.
  • The perfluoroelastomer O-rings are suitable for use with both aqueous and organic solutions.
A number of O-rings are used throughout the current Agilent Mark seven flame atomization system, and the earlier Agilent Mark VI flame atomization system, including on nebulizer components, the spray chamber and the burner. Regular inspection of the O-rings is recommended, especially when completing routine maintenance on the flame atomization system. O-rings should be replaced if they are perished, show excessive wear or have any nicks or cuts on the surface.
The regular black O-rings are suitable for use with aqueous solutions, including acid digests. For compatibility with organic solvents, perfluoroelastomer (white) O-rings should be fitted. The perfluoroelastomer O-rings are suitable for use with both aqueous and organic solutions.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty