Modern column technology based on superficially porous particles for reversed phase LC separations.
- Superficially porous, 'core-shell' particles that provide both improved throughput and higher resolution
- A scalable family of particles including 1.9, 2.7, and 4 μm for the best performance from your methods and instruments
- Superior peak shapes for faster, more accurate results due to high-purity silica and advanced bonding chemistries
- Easy method development with the selectivity from 12 reversed phase chemistries
- Excellent lot-to-lot reproducibility giving you confidence in your separation results
- Part of the InfinityLab family – designed for ease-of-use and maximum compatibility with Agilent instruments and supplies
- Long column life with uniquely robust 1.9 μm low-dispersion UHPLC columns
- Reduced need for sample preparation due to 2 μm frits on 2.7 and 4 μm columns
- UHPLC guard columns to further extend the life of your analytical column
- Enhanced usability, traceability, and security with a preprogrammed ID tag
Agilent InfinityLab Poroshell 120 superficially porous columns for reversed-phase LC separations offer exceptional efficiency and reliability. Eleven chemistries - from a C18 column to unique chemistries - are available in up to three particle diameters: 1.9 μm, 2.7 μm, and 4 μm. This range of chemistries provides a broad choice of selectivity for scalable LC method development, from traditional HPLC to UHPLC and to ultralow dispersion UHPLC.