Would you like to log in again?
Well Plate Tubes for Bond Elut NH2 Plates, Agilent Technologies
Supplier: Agilent Technologies
Bond Elut™
75505050 75505050
AG75505050PK 349 USD InStock
CAAG75505050 AG75505050
Well Plate Tubes for Bond Elut NH2 Plates, Agilent Technologies
Tubes Test Tubes
Bond Elut NH2 is a weak anion exchanger and a good choice for retention of very strong anions.
  • Streamlined sample preparation for lipid analysis
  • Features innovative EMR-Lipid sorbent technology
  • Time savings compared to traditional liquid-liquid extraction methods
  • mproved reproducibility compared to traditional liquid-liquid extraction methods, reducing the number of samples required for statistical evaluation
  • 2 ml well plate design is easily automatable for even more streamlined lipid analysis
Bond Elut NH2 is a weaker anion exchanger than sorbents such as SAX and is therefore a better choice for retention of very strong anions. Similar to Bond Elut Diol and SI sorbents, Bond Elut NH2 is excellent for the separation of structural isomers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty