Would you like to log in again?
Well Plate Tubes for Bond Elut C18 OH Plates, Agilent Technologies
Supplier: Agilent Technologies
7551601C 7551601C
AG7551601CPK 307 USD InStock
AG7551601C CAAG7551601C
Well Plate Tubes for Bond Elut C18 OH Plates, Agilent Technologies
Tubes Test Tubes
Bond Elut C18 OH enhances retention of basic compounds and promotes metabolite fractionation.
  • Silanol activity permits metabolite fractionation
  • Tight QC tolerances deliver batch-to-batch reproducibility
  • 50 Å pore size expands usage for higher molecular weight compounds
Bond Elut C18 OH is a nonendcapped version of the octadecyl bonded phases that enables the silanols on the silica surface to be more active. This low-load C18 phase has well-controlled silanol activity that permits the fractionation of metabolites and enhances retention of basic compounds compared to endcapped C18. The 150 Å pore size makes it a good choice for peptide extraction and higher molecular weight compounds.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty