Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 25, 2024. During this time, our site will be not available. We appreciate your patience.
Well Plate Tubes for Bond Elut Certify II Plates, Agilent Technologies
Supplier: Agilent Technologies
7551801C 7551801C
AG7551801CPK 341 USD InStock
CAAG7551801C AG7551801C
Well Plate Tubes for Bond Elut Certify II Plates, Agilent Technologies
Tubes Test Tubes
Bond Elut Certify II is a SAX/nonpolar SPE phase for extracting acidic analytes from biomatrices.
  • Ideal for nonpolar and anionic compounds
  • Optimized for acidic drug analysis
  • Bimodal, nonpolar and strong anion exchange
Bond Elut Certify II was developed specifically for the rapid extraction of acidic drugs and metabolites from urine and other biological matrices. Certify II is a mixed-mode cartridge packed with nonpolar C8 and SAX sorbent. It has been optimized for acidic drugs and other compounds that possess both nonpolar and anionic characteristics.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty