High-quality LC capillaries in a large variety of materials and dimensions.
- A variety of top-quality materials available such as stainless steel, titanium, PEEK-sleeved stainless steel, and PEEK-coated fused silica
- Agilent 0.6 mm od flexible 316 L grade stainless steel capillaries are corrosion resistant and much easier to handle than conventional rigid 1.6 mm od capillaries
- Color-coded for easy identification of inner diameters
- Bio-inert titanium or PEEK-sleeved stainless steel capillaries that fit perfectly for bio applications
All Agilent LC capillaries are engineered and manufactured to the highest quality standards to protect the integrity of your results at every step of your LC flow path. All capillaries are precision cut with square ends, are burr-free, have no inner-diameter distortion, and come in a variety of materials to suit your needs. Find the right capillaries quickly and easily using our online selection tool.