Would you like to log in again?
AdvanceBio RP-mAb, Agilent Technologies
Supplier: Agilent
793975-944 793975-904 785775-906 785975-906 783775-906 783775-906 785775-906 785975-906 799975-904 793975-944 795775-904 793975-904 795775-904 799975-904
AG795775-904EA 1059.36 USD InStock
CAAG793975-944 AG795775-904 CAAG799975-904 CAAG793975-904 CAAG785975-906 AG785775-906 AG783775-906 AG793975-904 CAAG795775-904 AG799975-904 CAAG783775-906 AG793975-944 AG785975-906 CAAG785775-906
AdvanceBio RP-mAb, Agilent Technologies
Chromatography Columns
Wide pore 450Å, superficially porous particles with C4, SB-C8 and Diphenyl ligands for intact proteins.
  • Improved accuracy: Superficially porous particles (3.5 µm) with wide pores (450Å) increase mAb resolution while maintaining compatibility with all LC instruments
  • Speed: Shorter analysis times compared to columns packed with fully porous particles of the same size
  • Lower costs: The robust Poroshell packed bed and 2 µm inlet frit extend column lifetime by helping prevent inlet blockage
  • Flexible method development: Range of chemistries - SB-C8, C4, and diphenyl
Agilent AdvanceBio RP-mAb columns are designed to optimize the performance of intact and reduced mAb analysis when analyzing monoclonal antibodies for biopharma discovery, development, and QA/QC applications. Analysis of intact and reduced monoclonal antibodies are critical measurements for characterizing therapeutic proteins and understanding their efficacy and stability. Poor chromatographic separations can result in rework and even compromise the accuracy of the characterization. Based on Poroshell technology with unique engineering for pore size and bonded phases, AdvanceBio RP-mAb columns deliver higher resolution and faster run times to provide accurate, reproducible results. Available in a range of chemistries – SB-C8, C4, and diphenyl. View our full range of bioHPLC columns for your area of interest, get resources, and take advantage of special offers.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty