Would you like to log in again?
Detector Lamps for Shimadzu LC Systems for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
8001-0702 8001-0704 8001-0702 8001-0701
101389-652EA 1161.54 USD InStock
CAAG8001-0702 101389-652 101389-658 AG8001-0702
Detector Lamps for Shimadzu LC Systems for HPLC, Agilent Technologies
Lights Deuterium Lamps
Lamps for selected Shimadzu HPLC systems, compatible with a range of detectors.
  • Deuterium lamps provide high output stability and intensity for extended detection capabilities and improved qualification at trace level
  • All deuterium lamps have a guaranteed lifetime of 2000 hours
  • Tungsten lamps offer coverage in the visible wavelength range for high-sensitivity detection
Agilent lamps for selected Shimadzu HPLC systems are designed and built to be compatible with a wide variety of detectors, including variable wavelength detectors (VWD), multiple wavelength detectors (MWD), diode array detectors (DAD), and photodiode array detectors (PDA). Both deuterium lamps and tungsten lamps are available.
All lamps adhere to the tightest specifications for consistent quality and reproducible performance over the entire lifetime of the lamp.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty