Would you like to log in again?
In-Line Filters for Shimadzu LC Systems, Agilent Technologies
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
8001-0808 8001-0808
AG8001-0808EA 171 USD InStock
AG8001-0808 CAAG8001-0808
In-Line Filters for Shimadzu LC Systems, Agilent Technologies
Filters In-Line Filters
Agilent offers in-line filters for selected Shimadzu HPLC systems made from clean, high-quality stainless steel parts, to help prevent time-wasting contamination and blockages.
  • In-line solvent filters, installed between the LC pump and injector, prevent blockages by removing particles from solvents before they reach the injector
  • Column inlet filters, positioned immediately before an LC column, remove particles from both the injection system and sample
Column inlet frit contamination can increase backpressure and reduce efficiency, especially in microbore columns.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty