Would you like to log in again?
Valve Replacement Parts for Shimadzu LC Systems, Agilent Technologies
Supplier: Agilent Technologies
8001-0614 8001-0809 8001-0603 8001-0603 8001-0809 8001-0601 8001-0601 8001-0614 8001-0608 8001-0608
AG8001-0608EA 251 USD InStock
CAAG8001-0608 AG8001-0608 CAAG8001-0614 AG8001-0809 CAAG8001-0603 CAAG8001-0601 AG8001-0601 AG8001-0603 CAAG8001-0809 AG8001-0614
Valve Replacement Parts for Shimadzu LC Systems, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
High-quality replacement valve parts for selected Shimadzu HPLC and LC/MS instruments.
  • PEEK offers high resistance, versatility, and applicability for the entire pH range between 0 and 14
  • Stators typically require replacement only if the ports or sealing surfaces become damaged, which can be prevented by using proper injection needles
Agilent valve supplies work seamlessly with selected Shimadzu HPLC systems for reliable, accurate, and reproducible results. Stators are available in 316 stainless steel, PEEK, and ceramic. Rotor seals are available in PEEK for a wide range of applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty