Would you like to log in again?
Valve Replacement Parts for Thermo/Dionex LC Systems, Agilent Technologies
Supplier: Agilent
8002-0860 8002-0807 8002-0602 8002-0603 8002-0603 8002-0602 8002-0860 8002-0812 8002-0812 8002-0807
AG8002-0860EA 112.94 USD InStock
AG8002-0860 AG8002-0807 CAAG8002-0602 CAAG8002-0860 AG8002-0603 AG8002-0812 CAAG8002-0807 AG8002-0602 CAAG8002-0603 CAAG8002-0812
Valve Replacement Parts for Thermo/Dionex LC Systems, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
A comprehensive range of replacement valve parts for selected Thermo Fisher Scientific HPLC systems.
  • Vespel is a polyimide with low wear and high chemical resistance recommended for use with solutions under pH 10
  • More basic solutions dissolve Vespel over time and damage the rotor seal
  • Tefzel is recommended for use in applications such as higher concentrations of methylene chloride or dimethyl sulfoxide (DMSO)
Agilent valve supplies work seamlessly with selected Thermo Fisher Scientific HPLC systems for reliable, accurate, and reproducible results. Stators are available in 316 stainless steel, PEEK, and ceramic depending on the specifications of the original equipment manufacturer. Rotor seals are available in Vespel and Tefzel for a wide range of applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty