Would you like to log in again?
Capillaries for Thermo/Dionex LC Systems, Agilent Technologies
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
8002-0824 8002-0823 8002-0808 8002-0819 8002-0808 8002-0822 8002-0825 8002-0818 8002-0803 8002-0823 8002-0821 8002-0818 8002-0825 8002-0822 8002-0826 8002-0819 8002-0824 8002-0820 8002-0821 8002-0826 8002-0820 8002-0803
AG8002-0818EA 152 USD InStock
CAAG8002-0820 CAAG8002-0821 CAAG8002-0822 CAAG8002-0818 CAAG8002-0808 CAAG8002-0819 CAAG8002-0823 CAAG8002-0824 CAAG8002-0825 CAAG8002-0803 CAAG8002-0826 AG8002-0818 AG8002-0819 AG8002-0808 AG8002-0803 AG8002-0825 AG8002-0826 AG8002-0823 AG8002-0824 AG8002-0821 AG8002-0822 AG8002-0820
Capillaries for Thermo/Dionex LC Systems, Agilent Technologies
Tubing Chromatography Tubing
Stainless steel capillaries and tubing for selected Thermo Fisher Scientific HPLC systems.
  • Made of ultra-clean stainless steel tubing
  • Ideal for most standard applications, and the best choice for reliable high-pressure sealing
  • Smooth inner surface for the lowest backpressure
  • Precision cutting for burr-free, square-cut ends without inner-diameter distortion
  • Both ends accept standard fittings
  • Predefined lengths for specific path locations to avoid peak broadening
  • Individually tested to ensure reproducibility
  • Available in non pre-swaged
Agilent offers a range of capillaries and tubing made from stainless steel for selected Thermo Fisher Scientific HPLC systems. Used with the right fittings, they are ideal for LC system plumbing, tight, leak-free connections, and zero dead volumes.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty