Would you like to log in again?
Nebulizers and Supplies for PerkinElmer ICP-OES Instruments, Agilent Technologies
Supplier: Agilent
8003-0327 8003-0327 G3139-68000 8003-0320 G3139-68000 8003-0320 8003-0974 8003-0974 8003-0326 8003-0326
AG8003-0326EA 1037.88 USD InStock
AG8003-0326 AG8003-0974 CAAG8003-0320 AG8003-0327 AGG3139-68000 CAAG8003-0326 CAAG8003-0974 AG8003-0320 CAAG8003-0327 CAAGG313968000
Nebulizers and Supplies for PerkinElmer ICP-OES Instruments, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Full portfolio of ICP-OES nebulizers provide optimal performance with your Perkin Elmer instrument.
  • Manufactured to fine tolerances for reproducible and stable sample uptake
  • Tight specifications deliver fine droplets for best signal intensity and optimum signal to noise
  • Fewer repeats means less downtime and improved productivity;Guaranteed to be compatible with PerkinElmer ICP-OES instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime;Qualified on a genuine PerkinElmer system
  • Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards, and services to ensure your PerkinElmer system runs at peak performance
  • Worldwide best-in-class technical support team to answer questions regarding part fit, function, or application-specific questions and concerns to help you get the most out of your PerkinElmer system
  • In stock and available worldwide for 24- to 48-hour delivery (in most regions)
  • Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies
  • Many parts in Agilent packaging are designed to be more durable, sturdy, and provide the best protection during shipping to ensure the part arrives intact
Agilent has a wide selection of Perkin Elmer supplies guarenteed to fit with your Perkin Elmer ICP-OES including the Agilent designed OneNeb 2 nebulizer. Agilent's nebulizers for PerkinElmer ICP-OES systems enhance performance productivity. Tight specifications for delivering fine droplets improve signal intensity and signal-to-noise. Choose from GemTip, GemCone, Meinhard, or inert MiraMist.
The Agilent OneNeb Series 2 nebulizer improves performance compared to glass concentric and inert types commonly used. The OneNeb Series 2 is up to four times more sensitive than most nebulizers and provides tolerance to high levels of dissolved solids and aggressive acids.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty