Would you like to log in again?
Torches and Supplies for PerkinElmer ICP-OES Instruments, Agilent Technologies
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
8003-0347 8003-0347 8003-0346 8003-0348 8003-0348 8003-0470 8003-0470 8003-0346 8003-0344 8003-0344
AG8003-0348EA 496 USD InStock
AG8003-0348 AG8003-0347 CAAG8003-0470 AG8003-0346 CAAG8003-0344 CAAG8003-0346 AG8003-0470 CAAG8003-0348 AG8003-0344 CAAG8003-0347
Torches and Supplies for PerkinElmer ICP-OES Instruments, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Perkin Elmer torches for Optima ICP-OES instruments.
  • Guaranteed to be compatible with PerkinElmer ICP-OES instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime;Qualified on a genuine PerkinElmer system
  • Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards, and services to ensure your PerkinElmer system runs at peak performance
  • Worldwide technical support team to answer questions regarding part fit, function, or application-specific questions and concerns to help you get the most out of your PerkinElmer system
  • In stock and available worldwide for 24- to 48-hour delivery (in most regions)
  • Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies
  • Many parts in Agilent packaging are designed to be more durable, sturdy, and provide the best protection during shipping to ensure the part arrives intact
Agilent torches for PerkinElmer ICP-OES are available in a wide range of styles from 0 to 3 slots, depending on the instrument and your application needs. Torches without slots are recommended for organic solvents. One-slot torches are the standard torch for aqueous solutions. Three-slot torches are typically used for organic analysis.
The Quick-Change module is useful when you are running different sample types and need separate sample introduction for each.The Quick-Connect torch module for PerkinElmer ICP-OES includes the torch, mounting block, 2 mm alumina injector, Scott spray chamber, and Cross-Flow nebulizer.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty