Would you like to log in again?
RF Load Coils and Igniter Supplies for PerkinElmer ICP-OES Instruments, Agilent Technologies
Supplier: Agilent Technologies
8003-0382 8003-0381 8003-0382 8003-0379 8003-0381 8003-0379
AG8003-0379EA 792 USD InStock
CAAG8003-0382 AG8003-0379 CAAG8003-0381 AG8003-0382 CAAG8003-0379 AG8003-0381
RF Load Coils and Igniter Supplies for PerkinElmer ICP-OES Instruments, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Perkin Elmer Optima ICP-OES Work/Load Coil and Ignitor Supplies.
  • Guaranteed to be compatible with PerkinElmer ICP-OES instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Qualified on a genuine PerkinElmer system
  • Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards, and services to ensure your PerkinElmer system runs at peak performance
  • Worldwide technical support team to answer questions regarding part fit, function, or application-specific questions and concerns to help you get the most out of your PerkinElmer system
  • In stock and available worldwide for 24- to 48-hour delivery (in most regions)
  • Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies
  • Many parts in Agilent packaging are designed to be more durable, sturdy, and provide the best protection during shipping to ensure the part arrives intact
Agilent RF Work/Load coils and igniter supplies are equivalent to the PerkinElmer ICP-OES versions for use with complete confidence.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty