Would you like to log in again?
Purge Extension Windows for PerkinElmer ICP-OES Instruments, Agilent Technologies
Supplier: Agilent
8003-0387 8003-0387 8003-0394 8003-0386 8003-0394 8003-0386 8003-0472 8003-0384 8003-0472 8003-0384
AG8003-0472EA 343.11 USD InStock
CAAG8003-0472 CAAG8003-0384 CAAG8003-0394 CAAG8003-0386 AG8003-0472 AG8003-0384 CAAG8003-0387 AG8003-0394 AG8003-0387 AG8003-0386
Purge Extension Windows for PerkinElmer ICP-OES Instruments, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Axial and Radial purge extension windows for Perkin Elmer Optima ICP-OES.
  • Guaranteed to be compatible with PerkinElmer ICP-OES instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Qualified on a genuine PerkinElmer system
  • Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards, and services to ensure your PerkinElmer system runs at peak performance
  • Worldwide technical support team to answer questions regarding part fit, function, or application-specific questions and concerns to help get the most out of PerkinElmer system
  • In stock and available worldwide for 24- to 48-hour delivery (in most regions)
  • Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies
  • Many parts in Agilent packaging are designed to be more durable, sturdy, and provide the best protection during shipping to ensure the part arrives intact
Purge extension windows for Perkin Elmer ICP-OES Optima instruments. Purge windows are available in both axial and radial and are guaranteed to work with your Perkin Elmer instrument. Agilent supplies are equivalent to PerkinElmer ICP-OES supplies for use with complete confidence.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty