Would you like to log in again?
Contact Cylinders for PerkinElmer GFAAS Systems, Agilent Technologies
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
8003-0957 8003-0949 8003-0401 8003-0401 8003-0949 8003-0957
AG8003-0401EA 399 USD InStock
CAAG8003-0957 AG8003-0401 AG8003-0957 CAAG8003-0949 AG8003-0949 CAAG8003-0401
Contact Cylinders for PerkinElmer GFAAS Systems, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Agilent contact cylinders equivalent to contact cylinders in PerkinElmer graphite furnace AAS system.
  • Guaranteed to be compatible with PerkinElmer graphite furnace AAS systems
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Qualified on a genuine PerkinElmer system;Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards, and services to ensure your PerkinElmer system runs at peak performance
  • Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies
  • Many parts in Agilent packaging are designed to be more durable, sturdy, and provide the best protection during shipping to ensure the part arrives intact
Agilent uses high-quality graphite and applies stringent quality controls to ensure contamination free and consistency of our contact cylinders for your PerkinElmer graphite furnace AA system. The standard THGA graphite contact cylinders, feature a modified injection port area that helps to minimize condensation and improve analytical performance with difficult matrices. The HGA contact cylinders have shaped graphite contacts to completely encase the graphite tube, ensuring the tube is positioned in a defined environment for uniform radiation conditions.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty