Would you like to log in again?
Graphite Tubes and Platforms for PerkinElmer GFAAS Systems, Agilent Technologies
Supplier: Agilent
8003-0410 8003-0416 8003-0421 8003-0959 8003-0958 8003-0959 8003-0954 8003-0421 8003-0954 8003-0416 8003-0410 8003-0958
AG8003-0416PK 664.92 USD InStock
CAAG8003-0959 CAAG8003-0958 AG8003-0416 AG8003-0954 CAAG8003-0421 CAAG8003-0410 AG8003-0959 AG8003-0958 AG8003-0410 AG8003-0421 CAAG8003-0416 CAAG8003-0954
Graphite Tubes and Platforms for PerkinElmer GFAAS Systems, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Agilent graphite tubes and L'vov platforms for PerkinElmer THGA and HGA graphite furnace AA systems.
  • Guaranteed to be compatible with PerkinElmer Graphite Furnace AAS systems
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Qualified on a genuine PerkinElmer system
  • Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards, and services to ensure your PerkinElmer system runs at peak performance
  • Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies
  • Many parts in Agilent packaging are designed to be more durable, sturdy, and provide the best protection during shipping to ensure the part arrives intact
Agilent graphite platforms and tubes for PerkinElmer THGA and HGA graphite furnace AA systems are batch tested to ensure they our demanding performance requirements for contamination, sensitivity, precision, electrical resistance, and lifetime. Partitioned tubes provide the best detection limits with samples that have low background. They are also ideal for organic solvents, and low viscosity samples that may spread and degrade precision. L'vov platforms isolate the sample from the tube walls to allow indirect heating of the sample on the platform, ensuring atomization occurs into a high temperature atmosphere, reducing interferences. The platforms also provide longer tube life through reduced attack by sample matrix and reagents.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty