Would you like to log in again?
Burners and Supplies for PerkinElmer AAS Instruments, Agilent Technologies
Supplier: Agilent
8003-0395 8003-0418 8003-0418 8003-0858 8003-0858 8003-0395
AG8003-0418EA 1798.68 USD InStock
CAAG8003-0418 CAAG8003-0858 CAAG8003-0395 AG8003-0418 AG8003-0858 AG8003-0395
Burners and Supplies for PerkinElmer AAS Instruments, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Agilent burner heads for PerkinElmer PinAAcle/AAnalyst flame atomic absorption systems.
  • Burner heads are designed to provide the best precision, efficient drainage, minimal burner blockage, and reduced interferences
  • Guaranteed to be compatible with PinAAcle/AAnalyst flame atomic absorption systems
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Qualified on a genuine PerkinElmer system
  • Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards, and services to ensure your PerkinElmer system runs at peak performance
  • Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies
  • Many parts in Agilent packaging are designed to be more durable, sturdy, and provide the best protection during shipping to ensure the part arrives intact
Agilent burner heads for PerkinElmer PinAAcle/AAnalyst flame AA systems are designed to provide the best precision, efficient drainage, minimal burner blockage, and reduced interferences. The 5 cm nitrous oxide burner head is required for nitrous oxide-acetylene operation, but can also be used with air-acetylene or rotated to provide reduced sensitivity. A 5 cm air-acetylene burner head is available for applications in which reduced sensitivity is required. It can be rotated to provide further sensitivity reduction, and has a wide slot to prevent clogging. The long pathlength 10 cm, single-slot air-acetylene burner head provides the best sensitivity for air-acetylene elements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty