Would you like to log in again?
Spray Chambers and Supplies for PerkinElmer ICP-MS Instruments, Agilent Technologies
Supplier: Agilent Technologies
8003-0946 8003-0588 8003-0948 8003-0524 8003-0514 8003-0588 8003-0947 8003-0947 8003-0946 8003-0948 8003-0514 8003-0524
AG8003-0524EA 109 USD InStock
CAAG8003-0946 AG8003-0524 AG8003-0588 CAAG8003-0948 CAAG8003-0947 AG8003-0946 AG8003-0514 CAAG8003-0588 AG8003-0948 AG8003-0947 CAAG8003-0524 CAAG8003-0514
Spray Chambers and Supplies for PerkinElmer ICP-MS Instruments, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Glass cyclonic, inert PFA spray chambers and supplies for PerkinElmer ELAN and NexION ICP-MS instuments.
  • Guaranteed to be compatible with PerkinElmer ICP-MS instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Qualified on a genuine PerkinElmer system
  • Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards, and services to ensure your PerkinElmer system runs at peak performance
  • Worldwide technical support team to answer questions regarding part fit, function, or application-specific questions and concerns, to help you get the most out of your PerkinElmer system
  • In stock and available worldwide for 24- to 48-hour delivery (in most regions)
  • Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies
  • Many parts in Agilent packaging are designed to be more durable, sturdy, and provide the best protection during shipping to ensure the part arrives intact
Agilent glass cyclonic, inert PFA spray chambers and supplies are equivalent to the PerkinElmer ICP-MS versions. These parts provide the best precision, efficient drainage, and reduced interferences and you can use them with complete confidence.
Use the single pass glass cyclonic spray chamber for good signal to noise performance with excellent washout. Alternatively, the double pass glass cyclonic spray chamber provides improved plasma stability with organic solvents and samples that have high levels of total dissolved solids.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty