Would you like to log in again?
Injectors and Injector Support Adaptors for PerkinElmer ICP-MS, Agilent Technologies
Supplier: Agilent Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
8003-0546 8003-0545 8003-0551 8003-0534 8003-0546 8003-0532 8003-0534 8003-0545 8003-0532 8003-0533 8003-0533 8003-0551
AG8003-0546EA 129 USD InStock
AG8003-0546 AG8003-0545 AG8003-0534 AG8003-0533 AG8003-0532 CAAG8003-0551 CAAG8003-0533 CAAG8003-0532 CAAG8003-0546 CAAG8003-0534 CAAG8003-0545 AG8003-0551
Injectors and Injector Support Adaptors for PerkinElmer ICP-MS, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Torch injectors and support adapters for PerkinElmer NexION 300/350 & ELAN 9000/6x00/DRCs ICP-MS.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Qualified on a genuine PerkinElmer system
  • Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards, and services to ensure your PerkinElmer system runs at peak performance
  • Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies
  • Many parts in Agilent packaging are designed to be more durable, sturdy, and provide the best protection during shipping to ensure the part arrives intact
Agilent quartz, sapphire, and alumina torch injectors, and injector support adapters for PerkinElmer NexION 300/350 & ELAN 9000/6x00/DRCs ICP-MS instruments. Achieve guaranteed performance with supplies for PE from Agilent that are equivalent to the PerkinElmer ICP-MS versions so you can use supplies with complete confidence.
Torch injectors are available in a wide range of types. Quartz injectors are recommended for volatile organic solvents and a wide range of non-HF applications. Sapphire injectors are recommended for environmental, geochemistry, and HF applications.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty