Would you like to log in again?
Torches and Supplies for PerkinElmer ICP-MS Instruments, Agilent Technologies
Supplier: Agilent Technologies
8003-0591 8003-0552 8003-0556 8003-0552 8003-0594 8003-0555 8003-0554 8003-0594 8003-0556 8003-0555 8003-0591 8003-0554
AG8003-0556EA 5672 USD InStock
AG8003-0556 CAAG8003-0591 AG8003-0555 AG8003-0554 CAAG8003-0594 CAAG8003-0552 CAAG8003-0555 AG8003-0591 CAAG8003-0554 AG8003-0552 CAAG8003-0556 AG8003-0594
Torches and Supplies for PerkinElmer ICP-MS Instruments, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Torches for the PerkinElmer ICP-MS systems.
  • Guaranteed to be compatible with PerkinElmer ICP-MS instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
  • Full portfolio of AA, ICP-OES, and ICP-MS consumables, standards and services to ensure your PerkinElmer system runs at peak performance
  • Worldwide technical support team to answer questions regarding part fit, function, or application-specific questions and concerns to help you get the most out of your PerkinElmer system
  • In stock and available worldwide for 24- to 48-hour delivery (in most regions)
  • Instruction guides and how-to videos available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies
Agilent torches and accessories are fully compatible with your PerkinElmer ICP-MS. The high efficiency demountable quartz torch operates at 25% reduced argon gas flow compared to the demountable quartz torch. The standard Agilent guarantee applies to our chemistries and supplies for PerkinElmer systems.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty